Learn More
Medchemexpress LLC Cefpodoxime Proxetil | 87239-81-4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN710110MG
Encompass_Preferred
Cefpodoxime Proxetil is an orally active, broad-spectrum third-generation cephalosporin with potent antibacterial activity against both Gram-positive and Gram-negative bacteria. It functions by binding to penicillin-binding proteins (PBPs), inhibiting peptidoglycan synthesis, and interfering with bacterial cell wall biosynthesis.
  • Orally active, broad-spectrum third-generation cephalosporin
  • Exhibits potent antibacterial activity against Gram-positive and Gram-negative bacteria
  • Inhibits peptidoglycan synthesis
  • Interferes with bacterial cell wall biosynthesis
  • Used to treat skin structure infections
  • Used to treat acute otitis media
  • Used to treat pharyngitis
  • Used to treat tonsillitis
  • Used to treat upper respiratory tract infections
  • Used to treat urinary tract infections
  • Used to treat sexually transmitted diseases
Catalog No. 50-004-19794