Learn More
Selleck Chemical LLC Cefpodoxime proxetil S5350-25mg
SDP
Supplier:  Selleck Chemical LLC S535025MG
Encompass_Preferred
Cefpodoxime Proxetil is a third generation semi-synthetic cephalosporin and a beta-lactam antibiotic with bactericidal activity
Catalog No. 50-310-8559