Learn More
Cayman Chemical CefprozIl-d4 5mg
SDP
Supplier:  Cayman Chemical 336395 MG
Encompass_Preferred
An internal standard for the quantification of cefprozil by GC- or LC-MS
Catalog No. 50-294-8966