Learn More
Medchemexpress LLC Cefprozil monohydrate | 121123-17-9 | 99.8% | 5 MG

Supplier: Medchemexpress LLC HYB04585MG
Cefprozil monohydrate is a second-generation cephalosporin antibiotic that exhibits broad-spectrum antibacterial activity and is orally active. It can be used for the study of pharyngitis, tonsillitis, otitis media, and uncomplicated skin infections.
- Molecular weight of 407.44
- Chemical formula C18H21N3O6S
- Appears as a white to off-white solid
- Inhibits various bacteria including S. pyogenes and E. coli
- Exhibits antibacterial activity in mouse models
- Orally active
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.