Learn More
Medchemexpress LLC Cefquinome (sulfate) | 118443-89-3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN6665100MG
Encompass_Preferred
Cefquinome sulfate is a broad-spectrum cephem antibiotic with inhibitory effects on various Gram-positive and Gram-negative bacteria, including Staphylococci, Streptococci, Pseudomonas, and Enterobacteriaceae. It functions by binding to penicillin-binding proteins (PBPs), thereby inhibiting bacterial cell wall synthesis. This product is for research use only.
  • Inhibits various Gram-positive and Gram-negative bacteria
  • Binds to penicillin-binding proteins (PBPs)
  • Inhibits bacterial cell wall synthesis
  • Demonstrates anti-infectious activity against Klebsiella pneumoniae
  • Reduces lung inflammatory cell infiltration and damage in models
  • Modulates inflammatory factors (TNF-α, IL-1β, IL-8, IL-4, IL-10, IL-13)
  • Appears as an off-white to light yellow solid
Catalog No. 50-004-20087