Learn More
Bioworld Cefsulodin Sodium Salt, 1 g
Supplier:  Bioworld 403100292
Encompass_Preferred
Third generation cephalosporin antibiotic with high specificity against Pseudomonas aeruginosa.A β lactam antibioticAppearance: SolidMelting Point: 175°C (lit.)(dec.)RTECS Number: UU1785000
Catalog No. 50-255-726