Learn More
Cayman Chemical CefsulodInsodIM salt hydra
SDP
Supplier:  Cayman Chemical 16127100 MG
Encompass_Preferred
A cephalosporin antibiotic; active against 45 isolates of P. aeruginosa (MIC90 = 3.1 UG/ml) and acts synergistically against P. aeruginosa isolates when combined with gentamicin, tobramycin, amikacin, or mezlocillin; binds to P. aeruginosa PBP1a, PBP1b, and PBP3 but not PBP2, PBP4, or PBP5; increases survival in mice infected with various P. aeruginosa strains (ED50s = 1.93-137 mg/kg for all)
Catalog No. 50-297-3143