Learn More
Cayman Chemical CeftazIdImehydrate 500mg
SDP
Supplier:  Cayman Chemical 14828500 MG
Encompass_Preferred
A cephalosporin antibiotic; active against isolates of numerous Gram-positive and Gram-negative bacteria, including E. coli, K. pneumoniae, P. mirabilis, and S. pyogenes (MICs = 0.015-8 UG/ml for all); binds to E. coli PBP1a, -1b, 2, and -3 (IC50s = 0.9, 3.4, 240, and 0.06 UG/ml, respectively); increases survival in mice infected with strains of S. aureus, E. coli, K. pneumoniae, Proteus, or Serratia (ED50s = 0.1-25 mg/kg for all)
Catalog No. 50-297-0693