Learn More
Medchemexpress LLC Ceftezole sodium | 41136-22-5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN70965MG
Encompass_Preferred
Ceftezole sodium is a broad-spectrum cephem antibiotic effective against many gram-positive and gram-negative bacteria. It also acts as an alpha-glucosidase inhibitor with in vivo anti-diabetic activity.
  • Broad-spectrum cephem antibiotic
  • Effective against many gram-positive and gram-negative bacteria
  • Alpha-glucosidase inhibitor
  • In vivo anti-diabetic activity
Catalog No. 50-004-19795