Learn More
Medchemexpress LLC Ceftezole sodium | 41136-22-5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN709650MG
Encompass_Preferred
Ceftezole sodium (CTZ sodium) is a broad-spectrum cephem antibiotic effective against many gram-positive and gram-negative bacteria. It also functions as an alpha-glucosidase inhibitor with in vivo anti-diabetic activity.
  • Broad-spectrum cephem antibiotic
  • Effective against many gram-positive and gram-negative bacteria
  • Functions as an alpha-glucosidase inhibitor
  • Exhibits in vivo anti-diabetic activity
  • For research use only
Catalog No. 50-004-17526