Learn More
Medchemexpress LLC Ceftibuten dihydrate | 118081-34-8 | 99.2% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0698A5MG
Encompass_Preferred
Ceftibuten dihydrate is an orally active cephalosporin antibiotic that demonstrates potent in vitro activity against a wide range of gram-negative and certain gram-positive pathogens. This biologically stable β-lactam antibiotic is transported by the small peptide transport system and is intended for research use only.
  • Orally active cephalosporin antibiotic
  • Potent activity against gram-negative and certain gram-positive pathogens
  • Highly active against *Haemophilus influenza*, *Escherichia coli*, *Klebsiella* sp., and *Proteus* sp.
  • Stable against most β-lactamase-producing organisms
  • Biologically stable β-lactam antibiotic
  • Transported by the small peptide transport system
  • For research use only
Catalog No. 50-004-08146