Learn More
Medchemexpress LLC Ceftibuten dihydrate | 118081-34-8 | 99.2% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0698A50MG
Encompass_Preferred
Ceftibuten dihydrate is an orally active cephalosporin antibiotic with potent in vitro activity against a wide range of gram-negative and certain gram-positive pathogens. It is stable in the presence of most β-lactamase-producing organisms, with the exception of Bacteroides fragilis.
  • Orally active cephalosporin antibiotic
  • Potent in vitro activity against a broad spectrum of gram-negative and some gram-positive pathogens
  • Highly active against Haemophilus influenza, Escherichia coli, Klebsiella sp., and Proteus sp.
  • Moderately active against Serratia sp. and Streptococcus pyogenes
  • Very active against strains of the family Enterobacteriaceae (mean MIC for 90% of strains = 0.25 μg/ml)
  • Stable in the presence of most β-lactamase-producing organisms, except Bacteroides fragilis
Catalog No. 50-004-05318