Learn More
Apexbio Technology LLC Ceftiofur HCl 103980-44-5 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B19075.1
Encompass_Preferred
Ceftiofur HCl (103980-44-5) is a small-molecule inhibitor targeting penicillin-binding proteins (PBPs) It is designed to inhibit bacterial cell wall synthesis thereby disrupting bacterial growth and structural integrity Ceftiofur HCl exerts its biological activity primarily through inhibition of peptidoglycan cross-linking in bacterial cell walls In in vitro studies Ceftiofur HCl demonstrates antibacterial activity with IC50 values in the range of approximately 0 5 5 g/mL depending on the bacterial species tested and experimental conditions Based on these pharmacological properties Ceftiofur HCl holds research potential in the investigation of bacterial resistance mechanisms antibiotic susceptibility assays and creation of analytical infection models
Catalog No. 50-000-92279