Learn More
Medchemexpress LLC Ceftobiprole | 209467-52-7 | 534.57 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1125791MG
Encompass_Preferred
Ceftobiprole (Ro 63-9141) is a broad-spectrum cephalosporin with high levels of in vitro activity against methicillin-resistant (MRSA) and vancomycin-resistant staphylococci (VRSA) and penicillin-resistant streptococci with a MIC90 value of 2 μg/mL for MRSA. It also inhibits gram-positive and gram-negative pathogens, and can be used for the study of hospital-acquired pneumonia (excluding ventilator-associated pneumonia) and community-acquired pneumonia.
  • Broad-spectrum cephalosporin
  • High levels of in vitro activity against methicillin-resistant (MRSA) and vancomycin-resistant staphylococci (VRSA) and penicillin-resistant streptococci
  • Inhibits gram-positive and gram-negative pathogens
  • Can be used for the study of hospital-acquired pneumonia and community-acquired pneumonia
Catalog No. 50-004-36352