Learn More
Medchemexpress LLC Ceftobiprole medocar 50mg | 252188-71-9 | 50MG
SDP
Supplier:  Medchemexpress LLC HY106574A50MG
Encompass_Preferred
Ceftobiprole medocaril (BAL5788) sodium is the parenteral proagent of Ceftobiprole (HY-112579) Ceftobiprole is a parenteral pyrrolidinone cephalosporin Ceftobiprole is a broad-spectrum cephalosporin with high levels of in vitro activity against methicillin- (MRSA) and vancomycin-resistant staphylococci (VRSA) and penicillin-resistant streptococci Ceftobiprole also inhibits gram-positive and gram-negative pathogens[1 [2
Catalog No. 50-002-42127