Learn More
Medchemexpress LLC Ceftolozane sulfate (CXA-101) | 936111-69-2 | 99.7% | 764.77 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY19806A10MM/1ML
Encompass_Preferred
Ceftolozane sulfate (CXA-101) is an antipseudomonal cephalosporin that demonstrates high affinity binding to *P. aeruginosa* essential PBPs (1b, 1c, 2 and 3). It functions by inhibiting cell wall synthesis through its binding to these PBPs. Ceftolozane sulfate effectively inhibits both *P. aeruginosa* and *Enterobacteriaceae*.
  • Antipseudomonal cephalosporin.
  • Binds to *P. aeruginosa* essential PBPs (1b, 1c, 2 and 3) with high affinity.
  • Inhibits cell wall synthesis.
  • Active against *P. aeruginosa* and *Enterobacteriaceae*.
Catalog No. 50-004-01710