Learn More
Medchemexpress LLC Ceftriaxone | 73384-59-5 | 99.6% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB07121MG
Encompass_Preferred
Ceftriaxone is a broad spectrum β-lactam third-generation cephalosporin antibiotic, demonstrating good antibacterial activity against various gram-negative and positive bacteria. It acts as a covalent inhibitor of GSK3β with an IC50 value of 0.78 μM and also inhibits Aurora B.
  • Exhibits anti-inflammatory activity
  • Exhibits antitumor activity
  • Exhibits antioxidant activity
  • Useful for studying bacterial infections
  • Useful for studying meningitis
Catalog No. 50-004-05253