Learn More
Medchemexpress LLC Ceftriaxone | 73384-59-5 | 99.6% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB07125MG
Encompass_Preferred
Ceftriaxone is a broad-spectrum β-lactam third-generation cephalosporin antibiotic. It exhibits good antibacterial activity against various gram-negative and positive bacteria. It is a covalent inhibitor of GSK3β and an inhibitor of Aurora B. Ceftriaxone also possesses anti-inflammatory, antitumor, and antioxidant activities, making it useful in studies of bacterial infections and meningitis.
  • Broad-spectrum β-lactam third-generation cephalosporin antibiotic
  • Good antibacterial activity against gram-negative and positive bacteria
  • Covalent inhibitor of GSK3β
  • Inhibitor of Aurora B
  • Exhibits anti-inflammatory properties
  • Exhibits antitumor properties
  • Exhibits antioxidant properties
  • Useful in studies of bacterial infections
  • Useful in studies of meningitis
Catalog No. 50-004-06848