Learn More
Medchemexpress LLC Ceftriaxone-d3 disodium | 1132650-38-4 | 96.6% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0712S1MG
Encompass_Preferred
Ceftriaxone-d3 (disodium) is the deuterium labeled version of Ceftriaxone, a third-generation cephalosporin antibiotic effective against many gram-negative and most gram-positive microorganisms. It also exhibits anti-inflammatory and antioxidant characteristics. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules to act as tracers for quantitation during drug development, and deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05254