Learn More
Research Products International Corp Ceftriaxone Disodium Salt Hemi (Heptahydrate), 1 G
SDP
Supplier:  Research Products International Corp C547001.0
Encompass_Preferred
Ceftriaxone Disodium Salt is a third-generation cephalosporin antibiotic. The hemi (heptahydrate) form indicates the presence of water molecules in the crystal structure. Ceftriaxone inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs), enzymes involved in the final stages of peptidoglycan synthesis. This interference leads to structural instability and eventual cell lysis. 
Broad-spectrum antibiotic effective against both Gram-positive and Gram-negative bacteria. May be used in microbiology research and studies focusing on antibacterial agents. Its inclusion in culture media can help selectively isolate bacteria that are susceptible to this antibiotic.
Catalog No. 50-197-7542