Learn More
Cayman Chemical CEHC Fatty Acids and Derivativ
SDP
Supplier:  Cayman Chemical 1000770610 MG
Encompass_Preferred
δ-CEHC is a major β-oxidation metabolite of δ-tocopherol.{4779,14205} Approximately 50% of a 3H-δ-tocopherol given as an intraperitoneal dose in rat is recovered in the urine as δ-CEHC, indicating this is the major route of metabolism.{14205}
Catalog No. 50-289-0449