Learn More
Apexbio Technology LLC Celastrol, 25mg. Cas: 34157-83-0 MFCD: MFCD03424073. Antioxidant, anti-inflammatory and immunosuppressive agent
SDP
Supplier:  Apexbio Technology LLC A260425
Encompass_Preferred
Celastrol is a potent proteasome inhibitor [1].Proteasomes are protein complexes that degrading unneeded or damaged proteins by proteolysis.Celastrol is a potent proteasome inhibitor and an antioxidant, anti-inflammatory and immunosuppressive agent. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4752