Learn More
AdipoGen Celastrol (50 mg)
SDP
Supplier:  AdipoGen AGCN20460M050
Encompass_Preferred
Chemical. CAS: 34157-83-0. Formula: C29H38O4. MW: 450.6. Isolated from Tripterygium wilfordii. Anti-obesity agent. Potent leptin sensitizer. Antioxidant. Potent lipid peroxidation inhibitor. NADPH oxidases (NOX1, 2, 4 and 5) inhibitor. Anti-inflammatory. NF-kappaB inhibitor. HSP90 inhibitor and HSP70 inducer. Anticancer agent. Apoptosis and autophagy inducer. Inhibits PI3K/Akt signaling pathways. Anti-metastatic and anti-angiogenic compound. Reduces VEGFR expression and inhibits HIF-1alpha pathway. Topoisomerase II inhibitor. 20S proteasome chymotrypsin-like activity inhibitor. Potential antiviral and anti-Alzheimer's agent.
Catalog No. 50-205-3633