Learn More
Medchemexpress LLC Celecoxib | 169590-42-5 | 99.9% | 5 G
SDP
Supplier:  Medchemexpress LLC HY143985G
Encompass_Preferred
Celecoxib is a selective and blood-brain barrier (BBB)-permeable non-steroidal anti-inflammatory drug (NSAID). It functions as a selective COX-2 inhibitor, demonstrating an IC50 of 40 nM.
  • It is a selective non-steroidal anti-inflammatory drug.
  • It can cross the blood-brain barrier.
  • It acts as a selective COX-2 inhibitor.
  • It has an IC50 of 40 nM for COX-2.
Catalog No. 50-003-55598