Learn More
TARGETMOL CHEMICALS INC Celecoxib 1G
SDP
Supplier:  TARGETMOL CHEMICALS INC T04661G
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Celecoxib (SC 58635) is a Nonsteroidal Anti-inflammatory Drug. The mechanism of action of celecoxib is as a Cyclooxygenase Inhibitor. Purity 99.98%
Catalog No. 50-288-0547