Learn More
Selleck Chemical LLC Celecoxib S1261-1g
SDP
Supplier:  Selleck Chemical LLC S12611G
Encompass_Preferred
Celecoxib is a selective COX-2 inhibitor with IC50 of 40 nM in Sf9 cells
Catalog No. 50-311-1070