Learn More
Medchemexpress LLC Celecoxib carboxylic acid | 170571-01-4 | MFCD09840210 | 98.3% | C17H12F3N3O4S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13423410MG
Encompass_Preferred
Celecoxib carboxylic acid is the carboxylic acid metabolite of celecoxib intended for use as a research reagent and reference standard. It is commonly used in drug metabolism studies, analytical method development, and metabolite profiling. The compound is reported with CAS 170571-01-4, molecular formula C17H12F3N3O4S, and a stated purity of 98.3%.
  • Metabolite reference for drug metabolism studies
  • Suitable for analytical method development and validation
  • Characterized by formula C17H12F3N3O4S for identification and tracking
  • High stated purity for reproducible experimental results
  • Available in multiple small-scale package sizes for laboratory use
Catalog No. 50-002-59466