Learn More
Medchemexpress LLC Celecoxib (Standard) | 169590-42-5 | 100.0% | C17H14F3N3O2S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY14398R
Encompass_Preferred
Celecoxib (Standard) is the analytical standard of Celecoxib, intended for research and analytical applications. It functions as a selective non-steroidal anti-inflammatory drug (NSAID) and a selective COX-2 inhibitor.
  • Analytical standard
  • Used as a reference standard for assays
  • IC50 of 40 nM for COX-2 inhibition
  • Commonly employed in qualitative, quantitative, and methodological research
  • Suitable for HPLC, GC, and MS experiments
Catalog No. 50-003-56893