Learn More
Medchemexpress LLC Celecoxib (Standard) | 169590-42-5 | 99.97% | C17H14F3N3O2S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14398R100MG
Encompass_Preferred
Celecoxib (Standard) is the analytical standard of Celecoxib, intended for research and analytical applications. It is a selective non-steroidal anti-inflammatory drug (NSAID) and a selective COX-2 inhibitor with an IC50 of 40 nM. This compound is an analytical standard used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard of Celecoxib
  • Intended for research and analytical applications
  • Selective non-steroidal anti-inflammatory drug (NSAID)
  • Selective COX-2 inhibitor with an IC50 of 40 nM
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-003-55599