Learn More
Medchemexpress LLC Celecoxib (Standard) | 169590-42-5 | 99.97% | C17H14F3N3O2S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14398R50MG
Encompass_Preferred
Celecoxib (Standard) is the analytical standard of Celecoxib, intended for research and analytical applications. Celecoxib itself is a selective non-steroidal anti-inflammatory drug (NSAID) and a selective COX-2 inhibitor with an IC50 of 40 nM. The compound is of analytical standard grade, serving as a reference standard for assays and commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for Celecoxib
  • Intended for research and analytical applications
  • Used as a reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-003-54430