Learn More
Regis Technologies Inc Celeris™ Amino Achiral SFC Column, 5 µm, 25 cm x 4.6 mm ID
SDP
Supplier:  Regis Technologies Inc 1790701300
Encompass
Achiral SFC Column. The Celeris™ Amino achiral SFC phase is a traditional propyl-amine phase bonded to 100A silica. This phase offers a high degree of polar selectivity over traditional silica or Ethyl Pyridine phases, allowing a high degree of retention for polar amine compounds.
Catalog No. 50-153-2975