Learn More
Selleck Chemical LLC Celiprolol hydrochloride
SDP
Supplier:  Selleck Chemical LLC S59232MG
Encompass_Preferred
Celiprolol is a 1-adrenoceptor antagonist with partial 2 agonist activity
Catalog No. 50-000-03939