Learn More
Beckman Coulter Beckman Coulter Cell Aligning/Removal Tool for Optima AUC Cells (362340)
Supplier:  Beckman Coulter 362340
Encompass
The Beckman Coulter Cell Aligning/Removal Tool (MPN 362340) is an accessory used with analytical ultracentrifuges to align sector cells before runs and assist with safe cell removal after runs. It is referenced as an accessory for Optima AUC instruments and is included in related starter and accessory kits.
  • Aligns analytical ultracentrifuge cells for correct positioning.
  • Assists safe removal of cells after runs to reduce handling damage.
  • Compatible with double-, six-, and eight-sector cells.
  • Included in Optima AUC starter and accessory kits.
  • Reduces risk of cell damage during handling.
Catalog No. NC2088384