Learn More
Eurofins Discoverx SPRINTer™ Human c-Myc Targeted Protein Degradation Cell-Based Assay Kit (K562)
Supplier:  Eurofins Discoverx 7911001C042
Encompass
The SPRINTer™ Human c-Myc Targeted Protein Degradation Stable Cell Line Assay (K562) contains a stable clonal cell line used for monitoring drug-induced protein turnover of c-Myc, supporting TPD studies. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56209