Learn More
Eurofins Discoverx SPRINTer™ Human IKZF2 Targeted Protein Degradation Cell-Based Assay Kit (Jurkat)
Supplier:  Eurofins Discoverx 7911010C19
Encompass
The SPRINTer™ Human IKZF2 Targeted Protein Degradation Stable Cell Line Assay (Jurkat) contains a stable clonal cell line used for monitoring drug-induced protein turnover of IKZF2, supporting TPD studies. This is a complete kit that includes the cell line vials, all required culture reagents for maintaining the cells in continuous culture, and detection reagents for running the assay.
Catalog No. 50-004-56216