Learn More
Eurofins Discoverx PathHunter Human AVPR1B β-Arrestin Cell-Based Assay Kit (CHO-K1)
Supplier:  Eurofins Discoverx 7930486C2
Encompass
The PathHunter Human AVPR1B β-Arrestin Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure AVPR1B (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55497