Learn More
Eurofins Discoverx PathHunter Mouse AVPR2 β-Arrestin Cell-Based Assay Kit (CHO-K1)
Supplier:  Eurofins Discoverx 7930596P2
Encompass
The PathHunter Mouse AVPR2 β-Arrestin Stable Cell Pool Assay (CHO-K1) contains a stable engineered cell pool used to measure mAVPR2 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell pool vials, all required culture reagents for maintaining the cells in continuous culture, and detection reagents for running the assay.
Catalog No. 50-004-55733