Learn More
Eurofins Discoverx PathHunter Mouse MC5R β-Arrestin Cell-Based Assay Kit (U2OS)
Supplier:  Eurofins Discoverx 7930609P3
Encompass
The PathHunter Mouse MC5R β-Arrestin Stable Cell Pine Assay (U2OS) contains a stable engineered cell pool used to measure mMC5R (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell pool vials, all required culture reagents for maintaining the cells in continuous culture, and detection reagents for running the assay.
Catalog No. 50-004-55734