Learn More
Eurofins Discoverx PathHunter Human MIP-3β (CCR7) Activated GPCR Internalization Cell-Based Assay Kit (U2OS)
Supplier:  Eurofins Discoverx 7930740C3
Encompass
The PathHunter Human MIP-3β (CCR7) Stable Cell Line Activated GPCR Internalization Assay (U2OS) contains a stable clonal cell line used to measure CCR7 (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56121