Learn More
Eurofins Discoverx PathHunter Human IL-10 (IL10RA/IL10RB) Dimerization Cell-Based Assay Kit (U2OS)
Supplier:  Eurofins Discoverx 7930985C3
Encompass
The PathHunter Human IL-10 (IL10RA/IL10RB) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL10RA/IL10RB. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55207