Learn More
Eurofins Discoverx PathHunter Human PD-1 (SHP1) Signaling Cell-Based Assay Kit (Jurkat)
Supplier:  Eurofins Discoverx 7931104C19
Encompass
The PathHunter Human PD-1 (SHP1) Signaling Stable Cell Line Assay (Jurkat) measures PD-1 signal by recruitment of SH2 domain protein to the activated receptor. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55950