Learn More
Eurofins Discoverx PathHunter Human PD-1 Signaling Reporter Cell-Based Assay Kit (Jurkat)
Supplier: Eurofins Discoverx 7931153C19
The PathHunter Human PD-1 Signaling NFAT Reporter Stable Cell Line Assay (Jurkat) pathway measures activation through detection of induced expression levels of a tagged reporter protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.