Learn More
Eurofins Discoverx PathHunter Human IL-3 (IL-3R (IL3RA/CSF2RB)) Signaling Reporter Cell-Based Assay Kit (U937)
Supplier:  Eurofins Discoverx 7931181C054
Encompass
The PathHunter Human IL-3 (IL-3R (IL3RA/CSF2RB)) Signaling STAT5 Reporter Stable Cell Line Assay (U937) monitor the activation of specific signaling pathways, utilizing the established PathHunter Enzyme Fragment Complementation (EFC) technology. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55972