Learn More
Eurofins Discoverx cAMP Hunter™ Human ADCYAP1R1 (PAC1) Cell-Based Assay Kit (CHO-K1)
Supplier: Eurofins Discoverx 7950064C2
The cAMP Hunter™ Human ADCYAP1R1 (PAC1) Gs Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure ADCYAP1R1 (GPCR) activity via increased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.