Learn More
Eurofins Discoverx cAMP Hunter™ Human OPRL1 (Opioid) Gi Cell-Based Assay Kit (CHO-K1)
Supplier:  Eurofins Discoverx 7950115C2
Encompass
The cAMP Hunter™ Human OPRL1 Gi Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure OPRL1 (GPCR) activity via decreased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55956