Learn More
Eurofins Discoverx BDKRB1 Gq Calcium Signaling Cell-Based Assay Kit (CHO-K1)
Supplier:  Eurofins Discoverx 7951011C2
Encompass
The BDKRB1 Gq Stable Cell Line Calcium Signaling Assay (CHO-K1) contains a stable clonal cell line used to measure BDKRB1 (GPCR) activation via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55981