Learn More
Eurofins Discoverx InCELL Hunter™ Human Bcl2L1-Bim Target Engagement Cell-Based Assay Kit (U2OS)
Supplier:  Eurofins Discoverx 7960037C3
Encompass
The InCELL Hunter™ Human BCL2L1-BIM Stable Cell Line Target Engagement Assay (U2OS) contains a stable clonal cell line used to measure protein binding via compound-induced binding partner disruption and degradation. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55392