Learn More
Eurofins Discoverx InCELL Hunter™ Human Hck Kinase Target Engagement Cell-Based Assay Kit (U2OS)
Supplier:  Eurofins Discoverx 7960041C15
Encompass
The InCELL Hunter™ Human HCK Stable Cell Line Target Engagement Assay (A549) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of HCK target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55387