Learn More
Eurofins Discoverx InCELL Hunter™ Human PI3K delta Kinase Target Engagement Cell-Based Assay Kit (HEK 293)
Supplier:  Eurofins Discoverx 7960061C1
Encompass
The InCELL Hunter™ Human PI3K delta Stable Cell Line Target Engagement Assay (HEK 293) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of PI3Kδ target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55322