Learn More
Eurofins Discoverx InCELL Hunter™ Human HPK1 Kinase Target Engagement Cell-Based Assay Kit (HEK 293)
Supplier: Eurofins Discoverx 7960083C1
The InCELL Hunter™ Human HPK1 Stable Cell Line Target Engagement Assay (HEK 293) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of HPK1 target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.